Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030253.3 | NP_084529.1 | 4 - 5 | 1575 | 95 | |
GenBank mRNA | AK037903.1 | - | 4 - 5 | 1142 | 95 | |
AK050032.1 | - | 4 - 5 | 1019 | 95 | ||
BC003281.1 | - | 4 - 5 | 1017 | 3581841 | 95 | |
BC010312.1 | - | 3 - 4 | 1000 | 95 | ||
BC070466.1 | - | 4 - 5 | 1575 | 30615225 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |