Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290797.1 | NP_001277726.1 | 21 - 22 | 2737 | 55 | |
NM_001290798.1 | NP_001277727.1 | 10 - 11 | 1466 | 55 | ||
NM_001290799.1 | NP_001277728.1 | 10 - 11 | 1463 | 55 | ||
NM_030266.4 | NP_084542.2 | 21 - 22 | 2623 | 55 | ||
NM_172971.3 | NP_766559.2 | 11 - 12 | 1499 | 55 | ||
GenBank mRNA | AF317838.1 | - | 19 - 20 | 2176 | 55 | |
AK035867.1 | - | 10 - 11 | 1467 | 55 | ||
AK149414.1 | - | 20 - 21 | 2530 | 55 | ||
BC120541.1 | - | 11 - 12 | 1495 | 40129464 | 55 | |
BC144747.1 | - | 11 - 12 | 1496 | 55 | ||
BC144748.1 | - | 10 - 11 | 1459 | 9053296 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |