Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134791.2 | NP_001128263.1 | 18 - 19 | 1789 | 97 | |
NM_133885.4 | NP_598646.4 | 21 - 22 | 2022 | 97 | ||
NM_173350.3 | NP_775485.2 | 19 - 20 | 1828 | 97 | ||
GenBank mRNA | AK035658.1 | - | 19 - 20 | 1778 | 97 | |
AK075981.1 | - | 8 - 9 | 808 | 97 | ||
AK131984.1 | - | 21 - 22 | 1994 | 97 | ||
AK172010.1 | - | 19 - 20 | 1815 | 97 | ||
BC002157.1 | - | 2 - 3 | 146 | 97 | ||
BC021507.1 | - | 17 - 18 | 1650 | 5347121 | 97 | |
BC023759.1 | - | 21 - 22 | 1977 | 97 | ||
BC026927.1 | - | 19 - 20 | 1711 | 5134780 | 97 | |
BC089163.1 | - | 18 - 19 | 1730 | 5367603 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |