Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145383.1 | NP_663358.1 | 1 - 2 | 437 | 86 | |
GenBank mRNA | AK044333.1 | - | 1 - 2 | 457 | 86 | |
AK044412.1 | - | 1 - 2 | 455 | 86 | ||
BC013125.1 | - | 1 - 2 | 437 | 4500760 | 86 | |
BC031766.1 | - | 1 - 2 | 438 | 4527040 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |