Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chtf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145409.2 | NP_663384.2 | 14 - 15 | 1861 | 71 | |
GenBank mRNA | AK052673.1 | - | 13 - 14 | 2072 | 71 | |
AK077947.1 | - | 14 - 15 | 1828 | 71 | ||
BC024142.1 | - | 14 - 15 | 1848 | 5149957 | 71 | |
BC083096.1 | - | 11 - 12 | 2579 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |