Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271358.1 | NP_001258287.1 | 6 - 7 | 568 | 119 | |
NM_001271359.1 | NP_001258288.1 | 6 - 7 | 568 | 119 | ||
NM_001271360.1 | NP_001258289.1 | 6 - 7 | 568 | 119 | ||
NM_145429.5 | NP_663404.1 | 6 - 7 | 568 | 119 | ||
GenBank mRNA | AK089542.1 | - | 6 - 7 | 503 | 119 | |
AK154874.1 | - | 6 - 7 | 488 | 119 | ||
AK159317.1 | - | 6 - 7 | 496 | 119 | ||
AK162889.1 | - | 6 - 7 | 505 | 119 | ||
AK170647.1 | - | 6 - 7 | 515 | 119 | ||
AK170889.1 | - | 6 - 7 | 478 | 119 | ||
BC016642.1 | - | 6 - 7 | 450 | 2651372 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |