Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010696.3 | NP_034826.2 | 17 - 18 | 1310 | 79 | |
GenBank mRNA | AK036727.1 | - | 17 - 18 | 1368 | 79 | |
AK037970.1 | - | 17 - 18 | 1344 | 79 | ||
AK088552.1 | - | 17 - 18 | 1432 | 79 | ||
AK153374.1 | - | 17 - 18 | 1315 | 79 | ||
AK168134.1 | - | 17 - 18 | 1325 | 79 | ||
AK170491.1 | - | 17 - 18 | 1587 | 79 | ||
BC006948.1 | - | 17 - 18 | 1310 | 3603181 | 79 | |
U20159.1 | - | 17 - 18 | 1349 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |