Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039482.1 | NP_001034571.1 | 8 - 9 | 1459 | 67 | |
GenBank mRNA | AK053102.1 | - | 8 - 9 | 1409 | 67 | |
AK220547.1 | - | 8 - 9 | 1438 | 67 | ||
BC019571.1 | - | 8 - 9 | 1431 | 4235202 | 67 | |
BC029801.1 | - | 2 - 3 | 182 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |