Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146008.2 | NP_666120.1 | 3 - 4 | 509 | 77 | |
GenBank mRNA | AK040850.1 | - | 3 - 4 | 560 | 77 | |
AK146035.1 | - | 3 - 4 | 834 | 77 | ||
BC008239.1 | - | 2 - 3 | 306 | 77 | ||
BC029663.1 | - | 3 - 4 | 509 | 5364434 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |