Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146036.1 | NP_666148.1 | 6 - 7 | 814 | 142 | |
GenBank mRNA | AK076069.1 | - | 6 - 7 | 824 | 142 | |
AK166644.1 | - | 6 - 7 | 787 | 142 | ||
BC023857.1 | - | 6 - 7 | 781 | 5346745 | 142 | |
BC025552.1 | - | 6 - 7 | 814 | 5320590 | 142 | |
BC036967.1 | - | 3 - 4 | 1382 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |