Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146152.3 | NP_666264.1 | 6 - 7 | 2020 | 84 | |
GenBank mRNA | AK052257.1 | - | 6 - 7 | 2021 | 84 | |
AK129200.1 | - | 6 - 7 | 1982 | 84 | ||
AK149978.1 | - | 6 - 7 | 2003 | 84 | ||
BC031823.1 | - | 6 - 7 | 1419 | 4188768 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |