Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177977.1 | NP_001171448.1 | 10 - 11 | 1490 | 81 | |
NM_001177978.1 | NP_001171449.1 | 10 - 11 | 1400 | 81 | ||
NM_146197.4 | NP_666309.1 | 10 - 11 | 1400 | 81 | ||
GenBank mRNA | AK057650.1 | - | 10 - 11 | 1400 | 81 | |
AK140827.1 | - | 10 - 11 | 1491 | 81 | ||
AK143757.1 | - | 10 - 11 | 1396 | 81 | ||
AK165353.1 | - | 10 - 11 | 1398 | 81 | ||
AK165401.1 | - | 10 - 11 | 1389 | 81 | ||
BC024424.1 | - | 1 - 2 | 102 | 81 | ||
BC031140.1 | - | 10 - 11 | 1376 | 4972673 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |