Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528913_s1
          See other OLFR1188 GE Assays ›
          Gene Symbol:
          Olfr1188
          Gene Name:
          olfactory receptor 1188
          Gene Aliases:
          MOR238-2
          Chromosome Location:
          Chr.2: 88556935 - 88560809 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528913_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1188
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146919.2 NP_667130.2 1 71 130
          GenBank mRNA BC104076.1 - 1 51 40041756 130
          BC104077.1 - 1 51 40041757 130

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1188

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334362 NM_146919.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334362 BC104076.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334362 BC104077.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334362 NM_146919.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334362 BC104076.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334362 BC104077.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164437 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334362 NM_146919.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334362 BC104076.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334362 BC104077.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334362 NM_146919.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334362 BC104076.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334362 BC104077.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334362 NM_146919.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334362 BC104076.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334362 BC104077.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334362 NM_146919.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334362 BC104076.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334362 BC104077.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410295 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164437 - -
          Whole Mouse Genome G4122A A_51_P334362 NM_146919.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334362 BC104076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334362 BC104077.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334362 NM_146919.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334362 BC104076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334362 BC104077.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108096 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164437 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01222958 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01222960 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01345151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01345158 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01345165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01805951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02095599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02273494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02125554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01345153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01670598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01353154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01302219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473601 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473600 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 684968 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2940463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221880 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221880 - -
          mouse-6 v1.1 (Illumina) scl4899.1.1_252-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4899.1.1_252-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221880 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221880 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl4899.1.1_252-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0