Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028031.3 | NP_082307.1 | 13 - 14 | 1495 | 85 | |
GenBank mRNA | AK010382.1 | - | 13 - 14 | 1495 | 85 | |
AK049640.1 | - | 12 - 13 | 1324 | 85 | ||
AK145855.1 | - | 13 - 14 | 1415 | 85 | ||
AK146196.1 | - | 13 - 14 | 1467 | 85 | ||
BC019493.1 | - | 13 - 14 | 1414 | 85 | ||
BC046599.1 | - | 13 - 14 | 1414 | 6311721 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |