Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smurf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038627.1 | NP_001033716.1 | 12 - 13 | 1650 | 72 | |
NM_029438.3 | NP_083714.3 | 12 - 13 | 1650 | 72 | ||
GenBank mRNA | AK015264.1 | - | 8 - 9 | 1049 | 72 | |
AK034736.1 | - | 3 - 4 | 248 | 72 | ||
AK154491.1 | - | 12 - 13 | 1651 | 72 | ||
BC029097.1 | - | 12 - 13 | 3660965 | 72 | ||
CB527254.1 | - | 4 - 5 | 574 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |