Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crispld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030209.3 | NP_084485.1 | 6 - 7 | 923 | 58 | |
GenBank mRNA | AK019034.1 | - | 6 - 7 | 919 | 58 | |
AK132171.1 | - | 6 - 7 | 866 | 58 | ||
AK149909.1 | - | 6 - 7 | 880 | 58 | ||
AK149925.1 | - | 6 - 7 | 880 | 58 | ||
AK150180.1 | - | 6 - 7 | 880 | 58 | ||
AK154308.1 | - | 6 - 7 | 923 | 58 | ||
BC090642.1 | - | 6 - 7 | 917 | 30653388 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |