Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahctf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026375.2 | NP_080651.2 | 12 - 13 | 2288 | 125 | |
GenBank mRNA | AB059278.1 | - | 12 - 13 | 1944 | 125 | |
BC138343.1 | - | 12 - 13 | 1931 | 8861363 | 125 | |
BC138344.1 | - | 12 - 13 | 1931 | 8861364 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |