Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100018219.mm.1 |
NM_026430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018219.mm.1 |
NM_026430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018219.mm.1 |
TC0100002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018220.mm.1 |
NM_026430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018220.mm.1 |
NM_026430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018220.mm.1 |
TC0100002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018215.mm.1 |
NM_026430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018215.mm.1 |
NM_026430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018215.mm.1 |
TC0100002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018217.mm.1 |
NM_026430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018217.mm.1 |
NM_026430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018217.mm.1 |
TC0100002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018221.mm.1 |
NM_026430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018221.mm.1 |
NM_026430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018221.mm.1 |
TC0100002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018210.mm.1 |
NM_026430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018210.mm.1 |
NM_026430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018210.mm.1 |
TC0100002266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018211.mm.1 |
NM_026430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018211.mm.1 |
NM_026430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018211.mm.1 |
TC0100002266.mm.1 |
MTA TC |