Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027326.3 | NP_081602.3 | 2 - 3 | 509 | 63 | |
GenBank mRNA | AF333960.1 | - | 1 - 2 | 170 | 63 | |
AK045331.1 | - | 2 - 3 | 363 | 63 | ||
BC021420.1 | - | 2 - 3 | 384 | 5012671 | 63 | |
CA328693.1 | - | 2 - 3 | 296 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |