Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242372.1 | NP_001229301.1 | 2878 | 126 | ||
NM_027389.3 | NP_081665.2 | 2827 | 126 | |||
GenBank mRNA | AK122531.1 | - | 2199 | 126 | ||
AK147575.1 | - | 2829 | 126 | |||
AY040842.1 | - | 790 | 126 | |||
BC108377.1 | - | 19 - 20 | 2118 | 126 |
Set Membership: |
Detect Genomic DNA Probe spans exons |