Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027592.3 | NP_081868.1 | 3 - 4 | 255 | 83 | |
GenBank mRNA | AK014903.1 | - | 2 - 3 | 177 | 83 | |
BC019453.1 | - | 3 - 4 | 206 | 4024616 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |