Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027810.3 | NP_082086.2 | 10 - 11 | 1132 | 105 | |
GenBank mRNA | AF521645.1 | - | 10 - 11 | 1131 | 105 | |
AK050291.1 | - | 10 - 11 | 1130 | 105 | ||
AK132661.1 | - | 10 - 11 | 1060 | 105 | ||
BC031505.1 | - | 10 - 11 | 1082 | 3494587 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |