Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172269.3 | NP_758473.3 | 3 - 4 | 625 | 64 | |
GenBank mRNA | AK029109.1 | - | 3 - 4 | 602 | 64 | |
AK033333.1 | - | 3 - 4 | 610 | 64 | ||
AK036915.1 | - | 3 - 4 | 610 | 64 | ||
AK039848.1 | - | 3 - 4 | 610 | 64 | ||
AK042471.1 | - | 3 - 4 | 613 | 64 | ||
AK044821.1 | - | 3 - 4 | 628 | 64 | ||
AK148077.1 | - | 3 - 4 | 614 | 64 | ||
AK160689.1 | - | 3 - 4 | 602 | 64 | ||
BC036129.1 | - | 3 - 4 | 553 | 4972693 | 64 | |
BC039176.1 | - | 3 - 4 | 532 | 4221232 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |