Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172557.2 | NP_766145.1 | 3 - 4 | 625 | 57 | |
GenBank mRNA | AJ250024.1 | - | 2 - 3 | 301 | 57 | |
AK046625.1 | - | 3 - 4 | 642 | 57 | ||
AK046633.1 | - | 3 - 4 | 626 | 57 | ||
AK051178.1 | - | 3 - 4 | 631 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |