Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3tc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172628.2 | NP_766216.2 | 9 - 10 | 1177 | 86 | |
GenBank mRNA | AK052534.1 | - | 9 - 10 | 1169 | 86 | |
AK143600.1 | - | 1178 | 86 | |||
AK173317.1 | - | 1 - 2 | 110 | 86 | ||
BC076633.1 | - | 2 - 3 | 252 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |