Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172736.3 | NP_766324.1 | 6 - 7 | 828 | 82 | |
GenBank mRNA | AK034310.1 | - | 6 - 7 | 833 | 82 | |
AK170629.1 | - | 6 - 7 | 836 | 82 | ||
BC041775.1 | - | 6 - 7 | 800 | 82 | ||
BC042658.1 | - | 6 - 7 | 816 | 5365041 | 82 | |
BC066768.1 | - | 6 - 7 | 819 | 6407146 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |