Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172757.3 | NP_766345.3 | 2 - 3 | 449 | 86 | |
GenBank mRNA | AK047782.1 | - | 2 - 3 | 480 | 86 | |
AK052635.1 | - | 2 - 3 | 476 | 86 | ||
AK134510.1 | - | 2 - 3 | 454 | 86 | ||
AK136424.1 | - | 2 - 3 | 459 | 87 | ||
AK140522.1 | - | 2 - 3 | 467 | 86 | ||
AK163715.1 | - | 2 - 3 | 436 | 86 | ||
BC065060.1 | - | 2 - 3 | 449 | 6402217 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |