Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172827.3 | NP_766415.1 | 3 - 4 | 1078 | 113 | |
GenBank mRNA | AK029094.1 | - | 3 - 4 | 1079 | 113 | |
AK036967.1 | - | 3 - 4 | 1079 | 113 | ||
AK161484.1 | - | 305 | 113 | |||
AK162332.1 | - | 955 | 113 | |||
BC066791.1 | - | 4 - 5 | 1402 | 30054751 | 113 | |
BC120925.1 | - | 3 - 4 | 1035 | 40098425 | 113 | |
BC120926.1 | - | 3 - 4 | 1035 | 40098428 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |