Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100011598.mm.1 |
NM_172843.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011598.mm.1 |
NM_172843.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011598.mm.1 |
TC0100001409.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_001160180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_001160180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_001160181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_001160181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_001160182.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_001160182.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_022329.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_022329.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_172843.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
NM_172843.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011591.mm.1 |
TC0100001409.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011596.mm.1 |
NM_172843.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011596.mm.1 |
NM_172843.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011596.mm.1 |
TC0100001409.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011597.mm.1 |
NM_172843.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011597.mm.1 |
NM_172843.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011597.mm.1 |
TC0100001409.mm.1 |
MTA TC |