Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172924.3 | NP_766512.2 | 3 - 4 | 335 | 103 | |
GenBank mRNA | AK048912.1 | - | 3 - 4 | 324 | 103 | |
AK090133.1 | - | 3 - 4 | 351 | 103 | ||
BC027695.1 | - | 1 - 2 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |