Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172992.3 | NP_766580.3 | 10 - 11 | 1429 | 94 | |
GenBank mRNA | AK037021.1 | - | 10 - 11 | 1316 | 94 | |
AK163376.1 | - | 10 - 11 | 1355 | 94 | ||
BC052841.1 | - | 10 - 11 | 1342 | 30050876 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |