Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173013.3 | NP_766601.2 | 4 - 5 | 482 | 98 | |
GenBank mRNA | AK032300.1 | - | 4 - 5 | 470 | 98 | |
AK082854.1 | - | 4 - 5 | 477 | 98 | ||
AK158860.1 | - | 2 - 3 | 251 | 98 | ||
AK206923.1 | - | 1 - 2 | 117 | 98 | ||
BC058166.1 | - | 4 - 5 | 463 | 98 | ||
BC079598.1 | - | 2 - 3 | 1843 | 98 | ||
BC150729.1 | - | 4 - 5 | 524 | 9087640 | 98 | |
DQ387862.1 | - | 4 - 5 | 470 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |