Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128170.2 | NP_001121642.1 | 6 - 7 | 1238 | 81 | |
NM_001128171.2 | NP_001121643.1 | 6 - 7 | 1363 | 81 | ||
NM_001276279.1 | NP_001263208.1 | 6 - 7 | 1360 | 81 | ||
NM_173369.3 | NP_775545.1 | 5 - 6 | 1229 | 81 | ||
GenBank mRNA | AK039054.1 | - | 7 - 8 | 1367 | 81 | |
AK122389.1 | - | 6 - 7 | 1344 | 81 | ||
AK147204.1 | - | 6 - 7 | 1207 | 81 | ||
AK147426.1 | - | 6 - 7 | 1202 | 81 | ||
AK167424.1 | - | 6 - 7 | 1324 | 81 | ||
BC042438.1 | - | 5 - 6 | 1182 | 3983771 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |