Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173400.2 | NP_775576.2 | 11 - 12 | 1495 | 108 | |
GenBank mRNA | AK036635.1 | - | 2 - 3 | 160 | 108 | |
AK051016.1 | - | 11 - 12 | 1516 | 108 | ||
AK077681.1 | - | 11 - 12 | 1517 | 108 | ||
AK136163.1 | - | 2 - 3 | 215 | 108 | ||
AK169222.1 | - | 11 - 12 | 1497 | 108 | ||
AK173206.1 | - | 11 - 12 | 1467 | 108 | ||
BC064725.1 | - | 6 - 7 | 692 | 108 | ||
BC068237.1 | - | 11 - 12 | 1495 | 30550558 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |