Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175484.3 | NP_780693.2 | 6 - 7 | 1143 | 89 | |
GenBank mRNA | AK047527.1 | - | 6 - 7 | 1085 | 89 | |
AK087414.1 | - | 6 - 7 | 1144 | 89 | ||
BC062649.1 | - | 6 - 7 | 875 | 6416110 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |