Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175543.3 | NP_780752.1 | 13 - 14 | 1711 | 95 | |
GenBank mRNA | AK046734.1 | - | 13 - 14 | 1713 | 95 | |
AK122559.1 | - | 5 - 6 | 547 | 95 | ||
AK147344.1 | - | 13 - 14 | 1907 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |