Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013867.2 | NP_038895.1 | 3 - 4 | 755 | 99 | |
GenBank mRNA | AF179566.1 | - | 3 - 4 | 703 | 99 | |
AK143894.1 | - | 3 - 4 | 543 | 99 | ||
AK158241.1 | - | 3 - 4 | 757 | 99 | ||
AK167578.1 | - | 3 - 4 | 574 | 99 | ||
BC023930.1 | - | 3 - 4 | 744 | 5324365 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |