Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024434.6 | NP_077754.3 | 7 - 8 | 945 | 93 | |
GenBank mRNA | AF334160.1 | - | 7 - 8 | 906 | 93 | |
AK002819.1 | - | 7 - 8 | 881 | 93 | ||
AK005334.1 | - | 7 - 8 | 858 | 93 | ||
AK008600.1 | - | 7 - 8 | 832 | 93 | ||
AK010384.1 | - | 7 - 8 | 829 | 93 | ||
AK010502.1 | - | 7 - 8 | 898 | 93 | ||
AK011604.1 | - | 7 - 8 | 831 | 93 | ||
AK166958.1 | - | 7 - 8 | 829 | 93 | ||
AK169032.1 | - | 7 - 8 | 831 | 93 | ||
AK186195.1 | - | 2 - 3 | 303 | 93 | ||
AK207598.1 | - | 2 - 3 | 279 | 93 | ||
BC016536.1 | - | 7 - 8 | 945 | 3488469 | 93 | |
CT010237.1 | - | 7 - 8 | 859 | 93 | ||
CT010301.1 | - | 7 - 8 | 859 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |