Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025287.2 | NP_079563.2 | 3 - 4 | 259 | 89 | |
GenBank mRNA | AB071989.1 | - | 1 - 2 | 68 | 89 | |
AK004669.1 | - | 3 - 4 | 89 | |||
AK028201.1 | - | 2 - 3 | 395 | 89 | ||
AK030746.1 | - | 2 - 3 | 446 | 89 | ||
AK132383.1 | - | 2 - 3 | 435 | 89 | ||
AK159482.1 | - | 2 - 3 | 412 | 89 | ||
AK166581.1 | - | 2 - 3 | 369 | 89 | ||
AK168185.1 | - | 2 - 3 | 381 | 89 | ||
AK168546.1 | - | 2 - 3 | 448 | 89 | ||
AY538613.1 | - | 1 - 2 | 71 | 89 | ||
BC043131.1 | - | 2 - 3 | 278 | 6811159 | 89 | |
BC045205.1 | - | 3 - 4 | 229 | 4988521 | 89 | |
BC064809.1 | - | 2 - 3 | 383 | 89 | ||
BY303667.1 | - | 3 - 4 | 265 | 89 | ||
BY726358.1 | - | 3 - 4 | 260 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |