Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080728.2 | NP_542766.1 | 9 - 10 | 877 | 69 | |
GenBank mRNA | AK040973.1 | - | 9 - 10 | 879 | 69 | |
AK147031.1 | - | 9 - 10 | 878 | 69 | ||
AK157742.1 | - | 9 - 10 | 878 | 69 | ||
AK169198.1 | - | 9 - 10 | 877 | 69 | ||
AK169371.1 | - | 9 - 10 | 877 | 69 | ||
AY056464.1 | - | 9 - 10 | 877 | 69 | ||
BC121789.1 | - | 8 - 9 | 833 | 40099448 | 69 | |
BC158018.1 | - | 954 | 9007324 | 69 | ||
BC158070.1 | - | 9 - 10 | 1132 | 9007368 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |