Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146145.2 | NP_666257.2 | 23 - 24 | 3568 | 78 | |
GenBank mRNA | AK133866.1 | - | 23 - 24 | 3512 | 78 | |
AK141210.1 | - | 23 - 24 | 3510 | 78 | ||
AK143650.1 | - | 23 - 24 | 3515 | 78 | ||
AK152128.1 | - | 23 - 24 | 3531 | 78 | ||
AK161759.1 | - | 1 - 2 | 1566 | 78 | ||
AK168058.1 | - | 3 - 4 | 386 | 78 | ||
BC024100.1 | - | 15 - 16 | 1978 | 78 | ||
BC031297.1 | - | 13 - 14 | 1689 | 78 | ||
S63728.1 | - | 23 - 24 | 3518 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |