Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023738.4 | NP_076227.1 | 21 - 22 | 2727 | 64 | |
GenBank mRNA | AF288694.1 | - | 16 - 17 | 1989 | 64 | |
AK004894.2 | - | 21 - 22 | 2728 | 64 | ||
AK054410.1 | - | 20 - 21 | 2819 | 64 | ||
AK089235.1 | - | 20 - 21 | 2620 | 64 | ||
AK129009.1 | - | 21 - 22 | 2637 | 64 | ||
AK170421.1 | - | 20 - 21 | 2805 | 64 | ||
AK170644.1 | - | 19 - 20 | 3186 | 64 | ||
AK172467.1 | - | 20 - 21 | 2937 | 64 | ||
BC022160.1 | - | 1 - 2 | 131 | 64 | ||
BC037693.1 | - | 19 - 20 | 2501 | 64 | ||
BC138843.1 | - | 21 - 22 | 2644 | 8861865 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |