Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175114.3 | NP_780323.2 | 7 - 8 | 911 | 64 | |
GenBank mRNA | AK029123.1 | - | 8 - 9 | 1309 | 64 | |
AK041436.1 | - | 8 - 9 | 1279 | 64 | ||
AK052264.1 | - | 8 - 9 | 1298 | 64 | ||
AK122426.1 | - | 8 - 9 | 1001 | 64 | ||
BC058971.1 | - | 7 - 8 | 853 | 6832507 | 64 | |
BC116262.1 | - | 7 - 8 | 878 | 64 | ||
BC116263.1 | - | 7 - 8 | 878 | 64 | ||
BC129961.1 | - | 7 - 8 | 878 | 64 | ||
BC129962.1 | - | 7 - 8 | 878 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |