Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289545.1 | NP_001276474.1 | 6 - 7 | 940 | 96 | |
NM_175183.5 | NP_780392.1 | 7 - 8 | 1119 | 96 | ||
GenBank mRNA | AK046849.1 | - | 7 - 8 | 1527 | 96 | |
AK131184.1 | - | 1 - 2 | 786 | 96 | ||
BC027136.1 | - | 3 - 4 | 1417 | 96 | ||
BC088996.1 | - | 5 - 6 | 714 | 96 | ||
BC094547.1 | - | 6 - 7 | 875 | 96 | ||
BC137599.1 | - | 6 - 7 | 861 | 8860618 | 96 | |
BC137600.1 | - | 6 - 7 | 888 | 8860619 | 96 | |
BC144756.1 | - | 6 - 7 | 868 | 9053304 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |