Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175358.4 | NP_780567.1 | 8 - 9 | 1000 | 98 | |
GenBank mRNA | AK029137.1 | - | 8 - 9 | 740 | 98 | |
AK032922.1 | - | 8 - 9 | 721 | 98 | ||
AK077949.1 | - | 8 - 9 | 749 | 98 | ||
AK132304.1 | - | 8 - 9 | 1002 | 98 | ||
AK160360.1 | - | 8 - 9 | 740 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |