Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175561.4 | NP_780770.2 | 23 - 24 | 4261 | 64 | |
GenBank mRNA | AK087907.1 | - | 8 - 9 | 1174 | 64 | |
AK220342.1 | - | 15 - 16 | 1823 | 64 | ||
BC068235.1 | - | 12 - 13 | 2328 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |