Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177324.2 | NP_796298.2 | 31 - 32 | 4382 | 61 | |
GenBank mRNA | AK029568.1 | - | 2 - 3 | 385 | 61 | |
BC015069.1 | - | 7 - 8 | 1098 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |