Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177381.3 | NP_796355.2 | 14 - 15 | 1649 | 71 | |
GenBank mRNA | AK140362.1 | - | 14 - 15 | 1649 | 71 | |
BC038030.1 | - | 15 - 16 | 1660 | 4020725 | 71 | |
BC048005.1 | - | 10 - 11 | 1004 | 71 | ||
CO432313.1 | - | 1 - 2 | 85 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |