Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178347.4 | NP_848124.1 | 5 - 6 | 552 | 88 | |
GenBank mRNA | AK031459.1 | - | 5 - 6 | 535 | 88 | |
AK038523.1 | - | 5 - 6 | 535 | 88 | ||
AK047789.1 | - | 5 - 6 | 535 | 88 | ||
AK048246.1 | - | 5 - 6 | 535 | 88 | ||
AK052567.1 | - | 5 - 6 | 552 | 88 | ||
AK075948.1 | - | 5 - 6 | 553 | 88 | ||
AK079681.1 | - | 4 - 5 | 620 | 88 | ||
AK081461.1 | - | 5 - 6 | 553 | 88 | ||
BC059013.1 | - | 5 - 6 | 521 | 6813730 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |