Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253719.1 | NP_001240648.1 | 9 - 10 | 1343 | 95 | |
NM_178673.4 | NP_848788.2 | 9 - 10 | 1603 | 95 | ||
GenBank mRNA | AF374460.1 | - | 9 - 10 | 1577 | 95 | |
AK033652.1 | - | 9 - 10 | 1594 | 95 | ||
AK046070.1 | - | 9 - 10 | 1344 | 95 | ||
AK081152.1 | - | 9 - 10 | 1594 | 95 | ||
AK138388.1 | - | 9 - 10 | 1895 | 95 | ||
BC075640.1 | - | 9 - 10 | 1566 | 30545529 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |